Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDRT1
Entrez Gene ID
694316
Gene Name
CMT1A duplicated region transcript 1
Gene Aliases
FBXW10
Chromosome Location
Chr.16: 15340325 - 15382565 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015118633.2 XP_014974119.2 - 1771 118
XM_015118634.2 XP_014974120.2 - 1771 118
XM_015118635.2 XP_014974121.2 - 1455 118
XM_015118636.2 XP_014974122.2 - 1171 118

View Full Product Details