Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGR2
Entrez Gene ID
709127
Gene Name
anterior gradient 2, protein disulphide isomerase family member
Gene Aliases
-
Chromosome Location
Chr.3: 98199805 - 98213070 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194304.2 NP_001181233.1 - 445 111
XM_015133907.2 XP_014989393.1 - 410 111

View Full Product Details