Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CGRRF1
Entrez Gene ID
695231
Gene Name
cell growth regulator with ring finger domain 1
Gene Aliases
-
Chromosome Location
Chr.7: 116622126 - 116650926 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266210.2 NP_001253139.1 - 731 147
XM_015143703.2 XP_014999189.1 - 700 147
GenBank mRNA JU321393.1 - - 733 147
JU472873.1 - - 738 147
JV045537.1 - - 748 147
JV639308.1 - - 758 147

View Full Product Details