Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAFD1
Entrez Gene ID
711635
Gene Name
TRAF-type zinc finger domain containing 1
Gene Aliases
-
Chromosome Location
Chr.11: 111824200 - 111853020 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266507.1 NP_001253436.1 - 2258 100
XM_015152832.2 XP_015008318.1 - 1813 100
XM_015152833.2 XP_015008319.1 - 2028 100
XM_028828950.1 XP_028684783.1 - 1864 100
GenBank mRNA JU323272.1 - - 2258 100
JU472769.1 - - 2258 100
JV044698.1 - - 1727 100
JV638460.1 - - 1769 100

View Full Product Details