Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPM3
Entrez Gene ID
712300
Gene Name
nucleophosmin/nucleoplasmin 3
Gene Aliases
-
Chromosome Location
Chr.9: 102275806 - 102277889 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194502.3 NP_001181431.2 - 136 136
GenBank mRNA JU327025.1 - - 113 136
JV049568.1 - - 117 136
JV565029.1 - - 124 136

View Full Product Details