Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS8
Entrez Gene ID
718962
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 8
Gene Aliases
-
Chromosome Location
Chr.14: 123053781 - 123077707 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015116124.2 XP_014971610.2 - 1965 135
GenBank mRNA JV569756.1 - - 1019 135
JV705819.1 - - 738 135

View Full Product Details