Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B4GALT7
Entrez Gene ID
701000
Gene Name
beta-1,4-galactosyltransferase 7
Gene Aliases
-
Chromosome Location
Chr.6: 175100935 - 175111609 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260676.1 NP_001247605.1 - 906 95
GenBank mRNA JU332772.1 - - 835 95
JU474183.1 - - 917 95
JV046085.1 - - 906 95
JV641307.1 - - 917 95

View Full Product Details