Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLL
Entrez Gene ID
711924
Gene Name
DNA polymerase lambda
Gene Aliases
EGK_19997
Chromosome Location
Chr.9: 102054715 - 102075556 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266906.2 NP_001253835.1 - 926 97
XM_015147969.2 XP_015003455.1 - 1060 97
XM_015147970.2 XP_015003456.1 - 1100 97
XM_015147971.2 XP_015003457.1 - 1097 97
XM_015147972.2 XP_015003458.1 - 1150 97
XM_015147973.2 XP_015003459.1 - 1098 97
XM_015147974.2 XP_015003460.1 - 960 97
XM_028853470.1 XP_028709303.1 - 972 97
XM_028853471.1 XP_028709304.1 - 1382 97
XM_028853473.1 XP_028709306.1 - 1097 97
XM_028853475.1 XP_028709308.1 - 1097 97
XM_028853478.1 XP_028709311.1 - 1097 97
XM_028853479.1 XP_028709312.1 - 1097 97
GenBank mRNA JU334040.1 - - 940 97
JV046782.1 - - 927 97
JV046783.1 - - 927 97
JV637757.1 - - 1236 97

View Full Product Details