Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX12
Entrez Gene ID
696408
Gene Name
sorting nexin 12
Gene Aliases
-
Chromosome Location
Chr.X: 68043380 - 68059481 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193757.3 NP_001180686.1 - 334 146
XM_001085637.4 XP_001085637.1 - 404 146
GenBank mRNA JU320897.1 - - 240 146
JU473715.1 - - 248 146
JV045049.1 - - 239 146
JV529101.1 - - 236 146

View Full Product Details