Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDIP1
Entrez Gene ID
705579
Gene Name
cell death inducing p53 target 1
Gene Aliases
C16orf5, C20H16orf5
Chromosome Location
Chr.20: 4593100 - 4622722 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194838.2 NP_001181767.1 - 263 137
XM_001097426.4 XP_001097426.1 - 425 137
XM_001097525.3 XP_001097525.1 - 290 137
XM_028840479.1 XP_028696312.1 - 288 137
XM_028840480.1 XP_028696313.1 - 369 137
XM_028840481.1 XP_028696314.1 - 398 137
GenBank mRNA JU321653.1 - - 287 137
JU471490.1 - - 273 137

View Full Product Details