Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COMMD9
Entrez Gene ID
716992
Gene Name
COMM domain containing 9
Gene Aliases
EGK_06347
Chromosome Location
Chr.14: 29174948 - 29188930 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266242.1 NP_001253171.1 - 498 108
XM_015114483.2 XP_014969969.1 - 474 108
XM_015114484.2 XP_014969970.1 - 376 108
GenBank mRNA JU326041.1 - - 430 108
JV045743.1 - - 498 108

View Full Product Details