Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- IMPA2
- Entrez Gene ID
-
704320
- Gene Name
- inositol monophosphatase 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.18: 71489063 - 71536737 on Build Mmul_10
- Gene Symbol
- LENG8
- Entrez Gene ID
-
695940
- Gene Name
- leukocyte receptor cluster member 8
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 54253459 - 54266107 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JV672174.1 |
- |
- |
238 |
148 |
View Full Product Details