Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MOK
Entrez Gene ID
106999576
Gene Name
MOK protein kinase
Gene Aliases
RAGE
Chromosome Location
Chr.7: 164454569 - 164540895 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015144514.2 XP_015000000.2 - 1214 133
XM_028851851.1 XP_028707684.1 - 1120 133
XM_028851852.1 XP_028707685.1 - 1117 133
XM_028851853.1 XP_028707686.1 - 1084 133
XM_028851855.1 XP_028707688.1 - 1211 133
XM_028851856.1 XP_028707689.1 - 1030 133
XM_028851857.1 XP_028707690.1 - 1027 133
XM_028851858.1 XP_028707691.1 - 1124 133
XM_028851859.1 XP_028707692.1 - 1121 133
XM_028851861.1 XP_028707694.1 - 1053 133
XM_028851862.1 XP_028707695.1 - 796 133
XM_028851863.1 XP_028707696.1 - 1050 133
XM_028851864.1 XP_028707697.1 - 467 133
XM_028851865.1 XP_028707698.1 - 492 133
XM_028851866.1 XP_028707699.1 - 417 133
XM_028851867.1 XP_028707700.1 - 750 133
XM_028851868.1 XP_028707701.1 - 1313 133
XM_028851870.1 XP_028707703.1 - 478 133
XM_028851871.1 XP_028707704.1 - 822 133
XM_028851872.1 XP_028707705.1 - 927 133

View Full Product Details