Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSD17B8
Entrez Gene ID
718119
Gene Name
hydroxysteroid 17-beta dehydrogenase 8
Gene Aliases
FabG-like
Chromosome Location
Chr.4: 136299942 - 136301916 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114958.1 NP_001108430.1 - 696 149
GenBank mRNA JU323170.1 - - 715 149
JU473200.1 - - 717 149
JV071116.1 - - 656 149

View Full Product Details