Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLA2G2E
Entrez Gene ID
704299
Gene Name
phospholipase A2 group IIE
Gene Aliases
-
Chromosome Location
Chr.1: 204809786 - 204813644 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001094364.2 XP_001094364.1 - 264 149

View Full Product Details