Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR1H
Entrez Gene ID
712151
Gene Name
RNA polymerase I subunit H
Gene Aliases
ZNRD1
Chromosome Location
Chr.4: 139825723 - 139829699 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015135848.2 XP_014991334.1 - 523 72
XM_015135849.2 XP_014991335.1 - 529 72
GenBank mRNA JU322719.1 - - 622 72
JU472298.1 - - 615 72
JV045782.1 - - 510 72

View Full Product Details