Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYGA
Entrez Gene ID
709748
Gene Name
crystallin gamma A
Gene Aliases
-
Chromosome Location
Chr.12: 95597828 - 95602371 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100820.1 NP_001094290.1 - 244 84
XM_015110803.2 XP_014966289.1 - 2030 84

View Full Product Details