Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JADE3
Entrez Gene ID
711390
Gene Name
jade family PHD finger 3
Gene Aliases
PHF16
Chromosome Location
Chr.X: 46874904 - 47025469 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001100171.4 XP_001100171.3 - 927 63
XM_002806209.3 XP_002806255.2 - 1167 63
XM_028842510.1 XP_028698343.1 - 1064 63
XM_028842511.1 XP_028698344.1 - 1038 63
GenBank mRNA JU337313.1 - - 939 63
JU478015.1 - - 869 63
JV048826.1 - - 1022 63
JV666670.1 - - 1025 63

View Full Product Details