Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NLGN4X
- Entrez Gene ID
-
106995148
- Gene Name
- neuroligin 4 X-linked
- Gene Aliases
-
NLGN4
- Chromosome Location
-
Chr.X: 5459345 - 5801293 on Build Mmul_10
- Gene Symbol
- NLGN4Y
- Entrez Gene ID
-
100270698
- Gene Name
- neuroligin 4, Y-linked
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Y: 4141220 - 4465753 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JU322425.1 |
- |
- |
696 |
91 |
View Full Product Details