Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JADE2
Entrez Gene ID
713338
Gene Name
jade family PHD finger 2
Gene Aliases
EGK_16861, PHF15
Chromosome Location
Chr.6: 131986624 - 132045049 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015140931.2 XP_014996417.1 - 379 90
XM_015140932.2 XP_014996418.1 - 380 90
XM_015140933.2 XP_014996419.1 - 382 90
XM_015140934.2 XP_014996420.1 - 383 90
XM_015140935.2 XP_014996421.1 - 511 90
XM_015140936.2 XP_014996422.1 - 403 90
XM_015140937.2 XP_014996423.1 - 236 90
XM_015140939.2 XP_014996425.1 - 744 90
XM_015140940.2 XP_014996426.1 - 272 90
XM_015140941.2 XP_014996427.1 - 511 90
GenBank mRNA JU333651.1 - - 249 90
JU474902.1 - - 168 90

View Full Product Details