The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_015109755.2 | XP_014965241.2 | - | 473 | 88 |
XM_015109757.2 | XP_014965243.2 | - | 564 | 88 | |
XM_028830252.1 | XP_028686085.1 | - | 442 | 88 | |
GenBank mRNA | JU322824.1 | - | - | 440 | 88 |
JV070530.1 | - | - | 438 | 88 |