Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM58
Entrez Gene ID
699182
Gene Name
tripartite motif containing 58
Gene Aliases
-
Chromosome Location
Chr.1: 1156804 - 1180524 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001086222.4 XP_001086222.2 - 923 113
XM_028848904.1 XP_028704737.1 - 901 113
XM_028848905.1 XP_028704738.1 - 656 113
GenBank mRNA JV647653.1 - - 300 113

View Full Product Details