Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AAR2
Entrez Gene ID
703053
Gene Name
AAR2 splicing factor
Gene Aliases
-
Chromosome Location
Chr.10: 71478779 - 71508577 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015149073.2 XP_015004559.2 - 1139 106
XM_015149074.2 XP_015004560.2 - 1198 106
XM_015149076.2 XP_015004562.2 - 1283 106
XM_028828405.1 XP_028684238.1 - 1387 106
GenBank mRNA JU322412.1 - - 862 106
JU470831.1 - - 807 106
JV044314.1 - - 851 106
JV567842.1 - - 912 106

View Full Product Details