Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOSIP
Entrez Gene ID
718888
Gene Name
nitric oxide synthase interacting protein
Gene Aliases
EGK_10894
Chromosome Location
Chr.19: 49522149 - 49548305 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194679.2 NP_001181608.2 - 836 98
XM_015124396.2 XP_014979882.2 - 850 98
GenBank mRNA JU322037.1 - - 732 98
JU471600.1 - - 736 98
JV452578.1 - - 727 98

View Full Product Details