Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAPPC12
Entrez Gene ID
707758
Gene Name
trafficking protein particle complex 12
Gene Aliases
TTC15
Chromosome Location
Chr.13: 105313693 - 105417791 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015111612.2 XP_014967098.2 - 1988 155
XM_015111613.2 XP_014967099.2 - 1949 155
XM_015111615.2 XP_014967101.2 - 1622 155
XM_028832011.1 XP_028687844.1 - 2054 155
GenBank mRNA JU325657.1 - - 1401 155
JV546840.1 - - 1416 155

View Full Product Details