Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAPPC4
Entrez Gene ID
702469
Gene Name
trafficking protein particle complex 4
Gene Aliases
-
Chromosome Location
Chr.14: 112084199 - 112089309 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257449.1 NP_001244378.1 - 647 153
GenBank mRNA JU320615.1 - - 647 153
JU473798.1 - - 602 153
JV043591.1 - - 696 153

View Full Product Details