Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOP16
Entrez Gene ID
696768
Gene Name
NOP16 nucleolar protein
Gene Aliases
EGK_17164
Chromosome Location
Chr.6: 173829726 - 173834018 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266227.1 NP_001253156.1 - 298 134
GenBank mRNA JV047311.1 - - 298 134

View Full Product Details