Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HACD3
Entrez Gene ID
709590
Gene Name
3-hydroxyacyl-CoA dehydratase 3
Gene Aliases
EGK_17601, PTPLAD1
Chromosome Location
Chr.7: 42080050 - 42129365 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266546.1 NP_001253475.1 - 280 104
XM_015142481.2 XP_014997967.1 - 64 104
GenBank mRNA JU322199.1 - - 275 104
JU332143.1 - - 275 104
JU332144.1 - - 275 104
JU473635.1 - - 280 104
JU473636.1 - - 280 104

View Full Product Details