Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM9
Entrez Gene ID
708473
Gene Name
transmembrane protein 9
Gene Aliases
EGK_05256
Chromosome Location
Chr.1: 58565084 - 58602074 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015120447.2 XP_014975933.1 - 307 116
XM_015120453.2 XP_014975939.1 - 310 116
XM_015120461.2 XP_014975947.1 - 595 116
XM_015120469.2 XP_014975955.1 - 291 116
XM_028851981.1 XP_028707814.1 - 427 116
XM_028851984.1 XP_028707817.1 - 399 116
GenBank mRNA JU333770.1 - - 199 116
JU333771.1 - - 190 116
JU472997.1 - - 164 116
JV046638.1 - - 240 116
JV637534.1 - - 171 116

View Full Product Details