Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC174
Entrez Gene ID
701654
Gene Name
coiled-coil domain containing 174
Gene Aliases
C11H3orf19, C3orf19, EGK_03990
Chromosome Location
Chr.2: 146005354 - 146026742 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193964.1 NP_001180893.1 - 125 99
GenBank mRNA JU334833.1 - - 106 99
JU475862.1 - - 111 99
JV047267.1 - - 100 99
JV653133.1 - - 102 99
JV716401.1 - - 111 99

View Full Product Details