Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PBDC1
Entrez Gene ID
705125
Gene Name
polysaccharide biosynthesis domain containing 1
Gene Aliases
CXHXorf26, CXorf26
Chromosome Location
Chr.X: 73017171 - 73022443 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194106.1 NP_001181035.1 - 288 131
XM_015127705.2 XP_014983191.1 - 363 131
GenBank mRNA JU322812.1 - - 188 131
JU475866.1 - - 188 131
JV508990.1 - - 191 131
JV653154.1 - - 210 131

View Full Product Details