Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTM
Entrez Gene ID
693345
Gene Name
neurotrimin
Gene Aliases
HNT
Chromosome Location
Chr.14: 123989722 - 124948076 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015116137.2 XP_014971623.1 - 893 122
XM_015116141.2 XP_014971627.1 - 893 122
XM_015116142.2 XP_014971628.1 - 893 122
XM_015116143.2 XP_014971629.1 - 695 122
XM_015116144.2 XP_014971630.1 - 695 122
XM_028834169.1 XP_028690002.1 - 801 122
XM_028834170.1 XP_028690003.1 - 1013 122
XM_028834171.1 XP_028690004.1 - 893 122
XM_028834172.1 XP_028690005.1 - 1013 122
XM_028834173.1 XP_028690006.1 - 1215 122
XM_028834174.1 XP_028690007.1 - 1013 122
GenBank mRNA JU334748.1 - - 628 122
JV600214.1 - - 1035 122

View Full Product Details