Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASL12
Entrez Gene ID
708527
Gene Name
RAS like family 12
Gene Aliases
-
Chromosome Location
Chr.7: 41589768 - 41604399 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265994.1 NP_001252923.1 - 645 73
XM_015142464.2 XP_014997950.1 - 831 73
GenBank mRNA JV047274.1 - - 645 73

View Full Product Details