Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM53C
Entrez Gene ID
714176
Gene Name
family with sequence similarity 53 member C
Gene Aliases
EGK_16899
Chromosome Location
Chr.6: 135742182 - 135755390 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266240.2 NP_001253169.1 - 324 100
XM_015140893.2 XP_014996379.1 - 1110 100
XM_028849274.1 XP_028705107.1 - 1335 100
XM_028849275.1 XP_028705108.1 - 3069 100
GenBank mRNA JU321320.1 - - 532 100
JU472890.1 - - 252 100
JV047281.1 - - 324 100

View Full Product Details