Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPG21
Entrez Gene ID
712815
Gene Name
SPG21 abhydrolase domain containing, maspardin
Gene Aliases
-
Chromosome Location
Chr.7: 41495380 - 41523700 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194890.2 NP_001181819.1 - 758 131
XM_001101919.4 XP_001101919.2 - 869 131
XM_015142466.2 XP_014997952.2 - 1194 131
XM_015142467.2 XP_014997953.1 - 790 131
XM_028850455.1 XP_028706288.1 - 1113 131
GenBank mRNA JU320616.1 - - 840 131
JU470837.1 - - 830 131
JV045126.1 - - 693 131

View Full Product Details