Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBX22
Entrez Gene ID
708338
Gene Name
T-box transcription factor 22
Gene Aliases
-
Chromosome Location
Chr.X: 76790586 - 76807593 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001102720.4 XP_001102720.2 - 340 73

View Full Product Details