Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL28
Entrez Gene ID
702075
Gene Name
kelch like family member 28
Gene Aliases
EGK_18149
Chromosome Location
Chr.7: 107218217 - 107338909 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JU322221.1 - - 110 107

View Full Product Details