Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAAF5
Entrez Gene ID
708567
Gene Name
dynein axonemal assembly factor 5
Gene Aliases
HEATR2
Chromosome Location
Chr.3: 33802789 - 33857765 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028845424.1 XP_028701257.1 - 1643 154

View Full Product Details