Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RMDN3
Entrez Gene ID
703902
Gene Name
regulator of microtubule dynamics 3
Gene Aliases
EGK_17370, FAM82A2
Chromosome Location
Chr.7: 17177274 - 17200389 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261760.1 NP_001248689.1 - 1293 131
XM_015141885.2 XP_014997371.2 - 2113 131
XM_015141886.2 XP_014997372.2 - 2108 131
XM_015141887.2 XP_014997373.1 - 1528 131
XM_028850306.1 XP_028706139.1 - 1397 131
GenBank mRNA JU345278.1 - - 1225 131
JU476222.1 - - 1293 131

View Full Product Details