Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM40
Entrez Gene ID
694585
Gene Name
transmembrane protein 40
Gene Aliases
-
Chromosome Location
Chr.2: 144113032 - 144137234 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028844475.1 XP_028700308.1 - 225 79

View Full Product Details