Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZGRF1
Entrez Gene ID
703497
Gene Name
zinc finger GRF-type containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 110821116 - 110906012 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015139049.2 XP_014994535.2 - 302 111
XM_015139050.2 XP_014994536.2 - 244 111
XM_015139051.2 XP_014994537.2 - 302 111
XM_015139053.2 XP_014994539.2 - 302 111
XM_015139054.2 XP_014994540.2 - 302 111
XM_015139055.2 XP_014994541.2 - 302 111

View Full Product Details