Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL26
Entrez Gene ID
718027
Gene Name
interleukin 26
Gene Aliases
-
Chromosome Location
Chr.11: 67731289 - 67759208 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001117154.4 XP_001117154.2 - 515 126

View Full Product Details