Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP41
Entrez Gene ID
703215
Gene Name
centrosomal protein 41
Gene Aliases
EGK_14139, TSGA14
Chromosome Location
Chr.3: 156689368 - 156741091 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266838.1 NP_001253767.1 - 78 89
GenBank mRNA JU321044.1 - - 78 89

View Full Product Details