Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ETAA1
Entrez Gene ID
697941
Gene Name
ETAA1 activator of ATR kinase
Gene Aliases
-
Chromosome Location
Chr.13: 41347996 - 41362947 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001093331.4 XP_001093331.2 - 414 150
XM_015112590.2 XP_014968076.2 - 209 150
GenBank mRNA JU335076.1 - - 461 150
JU476079.1 - - 454 150

View Full Product Details