Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASNSD1
Entrez Gene ID
709388
Gene Name
asparagine synthetase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.12: 76838466 - 76848210 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001106941.4 XP_001106941.2 - 234 145
GenBank mRNA JU323128.1 - - 197 145
JU472280.1 - - 207 145
JV047437.1 - - 98 145
JV703919.1 - - 245 145

View Full Product Details