Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLSCR3
Entrez Gene ID
721831
Gene Name
phospholipid scramblase 3
Gene Aliases
EGK_08121
Chromosome Location
Chr.16: 7155811 - 7161459 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265888.1 NP_001252817.1 - 982 82
XM_015118456.2 XP_014973942.1 - 1372 82
XM_015118457.2 XP_014973943.1 - 1376 82
XM_028835783.1 XP_028691616.1 - 1467 82
GenBank mRNA JU321589.1 - - 986 82
JV045213.1 - - 982 82

View Full Product Details