Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYZL6
Entrez Gene ID
716662
Gene Name
lysozyme like 6
Gene Aliases
EGK_08423
Chromosome Location
Chr.16: 32291127 - 32301164 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194655.1 NP_001181584.1 - 286 121
XM_015119036.2 XP_014974522.1 - 375 121

View Full Product Details