Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HCN3
Entrez Gene ID
717719
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 3
Gene Aliases
-
Chromosome Location
Chr.1: 95728654 - 95741224 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001115891.4 XP_001115891.1 - 406 121
GenBank mRNA JU490548.1 - - 128 121

View Full Product Details