Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC7A14
Entrez Gene ID
696121
Gene Name
solute carrier family 7 member 14
Gene Aliases
EGK_11907
Chromosome Location
Chr.2: 75380227 - 75503164 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257912.1 NP_001244841.1 - 592 87
XM_028843109.1 XP_028698942.1 - 1278 87
GenBank mRNA JU321422.1 - - 592 87

View Full Product Details