Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGF6
Entrez Gene ID
710739
Gene Name
fibroblast growth factor 6
Gene Aliases
-
Chromosome Location
Chr.11: 4522376 - 4545252 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001099809.3 XP_001099809.1 - 7635 146

View Full Product Details